Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172338.2 | NP_758841.1 | 2 | 222 | ||
GenBank mRNA | AK028692.1 | - | 2 | 227 | ||
AK051048.1 | - | 2 | 223 | |||
AK122407.1 | - | 469 | ||||
AK154946.1 | - | 243 | ||||
AK164023.1 | - | 2 | 238 |