Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122830.1 | NP_001116302.1 | 2 | 2395 | ||
NM_178771.3 | NP_848886.1 | 3 | 2578 | |||
GenBank mRNA | AK036321.1 | - | 3 | 2420 | ||
AK048601.1 | - | 3 | 2546 | |||
AK049946.1 | - | 3 | 2578 | |||
AK163170.1 | - | 3 | 2578 | |||
AK178365.1 | - | 1 | 371 | |||
AK211135.1 | - | 1 | 100 | |||
BC023909.1 | - | 3 | 2593 | 5322205 | ||
BC025484.1 | - | 3 | 2495 | 5151231 | ||
BC098185.1 | - | 3 | 2559 | 3594528 |