Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172117.5 | NP_742115.2 | 4 | 580 | ||
GenBank mRNA | AK030789.1 | - | 4 | 581 | ||
AK156685.1 | - | 3 | 495 | |||
BC028318.1 | - | 4 | 524 | |||
BC038126.1 | - | 4 | 569 | 1348160 |