Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172203.2 | NP_757340.1 | 10 | 1321 | ||
GenBank mRNA | AF539799.1 | - | 10 | 1175 | ||
AK136432.1 | - | 10 | 1236 | |||
AY174116.1 | - | 7 | 916 |