Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017.2 | NP_001008.1 | 4, 5 | 361 | ||
GenBank mRNA | AK312060.1 | - | 4, 5 | 355 | ||
BC000475.1 | - | 1 | 340 | 2961637 | ||
BC006772.1 | - | 1 | 342 | 2899987 | ||
BC029732.1 | - | 4, 5 | 341 | 4388273 | ||
BC066322.1 | - | 4, 5 | 355 | 4816284 | ||
BC100032.1 | - | 4, 5 | 341 | 6025884 | ||
L01124.1 | - | 4, 5 | 361 | |||
L05090.1 | - | 3, 4 | 297 | |||
X79239.1 | - | 4, 5 | 329 |