Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172260.3 | NP_758464.2 | 7 | 3086 | ||
GenBank mRNA | AK031622.1 | - | 7 | 3083 | ||
AK140820.1 | - | 7 | 3087 | |||
AK148267.1 | - | 5 | 2226 |