Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172269.3 | NP_758473.3 | 4 | 805 | ||
GenBank mRNA | AK029109.1 | - | 4 | 782 | ||
AK033333.1 | - | 4 | 790 | |||
AK036915.1 | - | 4 | 790 | |||
AK039848.1 | - | 4 | 790 | |||
AK042471.1 | - | 4 | 793 | |||
AK044821.1 | - | 4 | 808 | |||
AK148077.1 | - | 4 | 794 | |||
AK160689.1 | - | 4 | 782 | |||
BC036129.1 | - | 4 | 733 | 4972693 | ||
BC039176.1 | - | 4 | 712 | 4221232 |