Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172282.2 | NP_758486.1 | 5 | 1382 | ||
GenBank mRNA | AK033499.1 | - | 5 | 1143 | ||
AK046065.1 | - | 5 | 1172 | |||
AK049974.1 | - | 5 | 1141 | |||
AK088866.1 | - | 5 | 1166 | |||
AK148252.1 | - | 5 | 1185 | |||
BC039955.1 | - | 5 | 1128 | 4236297 |