Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pced1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172293.4 | NP_758497.1 | 6 | 1708 | ||
GenBank mRNA | AK051480.1 | - | 6 | 1709 | ||
BC038822.1 | - | 2 | 1317 | 4168445 | ||
BC086317.1 | - | 1 | 1146 | 30429924 |