Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290419.1 | NP_001277348.1 | 24 | 4470 | ||
NM_172339.4 | NP_758842.1 | 23 | 4351 | |||
GenBank mRNA | AK077522.1 | - | 23 | 4318 | ||
BC044754.2 | - | 8 | 2768 | |||
BC049809.1 | - | 1 | 3471 | |||
BC057031.1 | - | 24 | 4436 | 6833968 |