Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083331.2 | NP_001076800.1 | 1 | 117 | ||
NM_001276406.1 | NP_001263335.1 | 1 | 117 | |||
NM_172394.3 | NP_765982.2 | 1 | 117 | |||
GenBank mRNA | AK002643.1 | - | 1 | 104 | ||
AK028563.1 | - | 1 | 107 | |||
AK039007.1 | - | 1 | 107 | |||
AK050893.1 | - | 1 | 106 | |||
AK085776.1 | - | 1 | 118 | |||
AK139738.1 | - | 1 | 25 | |||
AK151739.1 | - | 1 | 107 | |||
AK160619.1 | - | 1 | 118 | |||
AK166214.1 | - | 1 | 107 | |||
BC032929.1 | - | 1 | 98 | 1379673 |