Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ostm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172416.3 | NP_766004.1 | 1 | 253 | ||
GenBank mRNA | AF533890.1 | - | 1 | 252 | ||
AK004546.1 | - | 1 | 230 | |||
AK044897.1 | - | 1 | 252 | |||
AK050513.1 | - | 1 | 227 | |||
AK053664.1 | - | 1 | 203 | |||
BC057635.1 | - | 1 | 191 | 4910872 |