Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001041.3 | NP_001032.2 | 33 | 3975 | ||
XM_011513078.2 | XP_011511380.1 | 33 | 3979 | |||
GenBank mRNA | BC115034.1 | - | 33 | 3988 | 7962025 | |
BC116452.1 | - | 33 | 3988 | 7962021 | ||
BC116453.1 | - | 33 | 3988 | |||
BC132834.1 | - | 33 | 3968 | 40146856 | ||
BC132860.1 | - | 33 | 3968 | 40146882 | ||
X63597.1 | - | 33 | 3974 |