Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thoc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172438.3 | NP_766026.1 | 2, 3 | 160 | ||
GenBank mRNA | AK050734.1 | - | 2, 3 | 160 | ||
BC020332.1 | - | 1, 2 | 125 | |||
BC021779.1 | - | 1, 2 | 134 | |||
BC039758.1 | - | 2, 3 | 161 | 3995737 |