Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172448.3 | NP_766036.2 | 5 | 1168 | ||
GenBank mRNA | AK028750.1 | - | 4 | 527 | ||
AK032782.1 | - | 4 | 1211 | |||
AK153638.1 | - | 4 | 2446 | |||
BC075707.1 | - | 3 | 304 | |||
BC131957.1 | - | 3 | 286 | 40130234 |