Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Panx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172454.2 | NP_766042.2 | 1, 2 | 343 | ||
GenBank mRNA | AK028288.1 | - | 1, 2 | 310 | ||
AK036933.1 | - | 1, 2 | 299 | |||
BB849008.1 | - | 1, 2 | 358 | |||
BC052822.1 | - | 1, 2 | 311 | 30058169 |