Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172461.3 | NP_766049.2 | 2, 3 | 407 | ||
GenBank mRNA | AK030042.1 | - | 2, 3 | 366 | ||
AK030186.1 | - | 2, 3 | 410 | |||
AK043543.1 | - | 2, 3 | 646 | |||
AK054237.1 | - | 2, 3 | 301 | |||
BC119588.1 | - | 1, 2 | 241 | 40058889 | ||
BC119589.1 | - | 1, 2 | 241 | 40058890 |