Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172466.2 | NP_766054.2 | 4 | 985 | ||
GenBank mRNA | AK035797.1 | - | 4 | 885 | ||
AK036137.1 | - | 2 | 151 | |||
AK053851.1 | - | 3 | 1093 | |||
AK140876.1 | - | 4 | 761 | |||
BC094674.1 | - | 4 | 640 | 30548647 | ||
BC144816.1 | - | 2 | 243 | |||
BC144817.1 | - | 2 | 242 | |||
CB246179.1 | - | 3 | 634 |