Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105196.1 | NP_001098666.1 | 3, 4 | 810 | ||
NM_001105197.1 | NP_001098667.1 | 3, 4 | 496 | |||
NM_001271489.1 | NP_001258418.1 | 3, 4 | 791 | |||
NM_001271490.1 | NP_001258419.1 | 3, 4 | 963 | |||
NM_001271491.1 | NP_001258420.1 | 3, 4 | 530 | |||
NM_172472.3 | NP_766060.2 | 3, 4 | 810 | |||
GenBank mRNA | AK089762.1 | - | 2, 3 | 572 | ||
AK139756.1 | - | 3, 4 | 807 | |||
AK144657.1 | - | 3, 4 | 775 | |||
AK144751.1 | - | 3, 4 | 778 | |||
AK145029.1 | - | 3, 4 | 930 | |||
AK145832.1 | - | 3, 4 | 775 | |||
AK147405.1 | - | 2, 3 | 1265 | |||
AK150372.1 | - | 3, 4 | 562 | |||
AK151447.1 | - | 3, 4 | 481 | |||
AK152221.1 | - | 3, 4 | 481 | |||
AK153051.1 | - | 3, 4 | 562 | |||
AK155883.1 | - | 3, 4 | 532 | |||
AK159162.1 | - | 3, 4 | 691 | |||
AK181999.1 | - | 1, 2 | 89 | |||
AK185744.1 | - | 1, 2 | 89 | |||
BC056358.1 | - | 3, 4 | 798 | 6813486 | ||
BC063047.1 | - | 3, 4 | 798 | 6813486 | ||
S76673.1 | - | 1, 2 | 148 |