Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172479.3 | NP_766067.2 | 3 | 247 | ||
GenBank mRNA | AK054485.1 | - | 3 | 234 | ||
AK077667.1 | - | 3 | 216 | |||
AK155928.1 | - | 3 | 200 | |||
AK158125.1 | - | 2 | 368 | |||
BC152401.1 | - | 3 | 216 | 40057282 | ||
BY247469.1 | - | 3 | 237 |