Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001062.3 | NP_001053.2 | 6 | 941 | ||
GenBank mRNA | AK292990.1 | - | 6 | 858 | ||
BM769693.1 | - | 2 | 187 |