Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thsd7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172485.3 | NP_766073.2 | 2 | 445 | ||
GenBank mRNA | AK051714.1 | - | 2 | 287 | ||
AK052119.1 | - | 2 | 448 | |||
AK083798.1 | - | 2 | 273 | |||
BB658510.1 | - | 2 | 445 | |||
BC063250.1 | - | 2 | 275 | 6410178 |