Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poglut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300827.1 | NP_001287756.1 | 3 | 521 | ||
NM_172380.4 | NP_759012.1 | 3 | 318 | |||
GenBank mRNA | AK031608.1 | - | 3 | 257 | ||
AK035948.1 | - | 3 | 266 | |||
AK036224.1 | - | 3 | 248 | |||
AK041321.1 | - | 3 | 256 | |||
AK045278.1 | - | 3 | 466 | |||
AK079886.1 | - | 3 | 287 | |||
AK082556.1 | - | 3 | 488 | |||
AK167641.1 | - | 3 | 488 | |||
AK197458.1 | - | 2 | 358 | |||
AK215456.1 | - | 3 | 160 | |||
AK216766.1 | - | 3 | 160 | |||
BC026809.1 | - | 3 | 269 | 4205469 | ||
CB249259.1 | - | 3 | 319 |