Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172404.2 | NP_765992.2 | 5, 6 | 627 | ||
GenBank mRNA | AK008165.1 | - | 3, 4 | 404 | ||
AK031511.1 | - | 5, 6 | 640 | |||
AK042391.1 | - | 4, 5 | 573 | |||
AK088404.1 | - | 4, 5 | 535 | |||
AK163165.1 | - | 4, 5 | 436 | |||
BC016206.1 | - | 5, 6 | 729 | 4912335 | ||
BC025124.1 | - | 4, 5 | 760 | 4486058 | ||
BC052047.1 | - | 5, 6 | 627 | 5687475 |