Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172412.2 | NP_766000.1 | 2 | 367 | ||
GenBank mRNA | AK010515.1 | - | 2 | 397 | ||
AK048605.1 | - | 2 | 393 | |||
AK049639.1 | - | 2 | 368 | |||
AK160959.1 | - | 2 | 413 | |||
BC056941.1 | - | 1 | 1066 | |||
BC083180.1 | - | 2 | 374 | 5707617 |