Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med13l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172424.4 | NP_766012.3 | 2, 3 | 384 | ||
GenBank mRNA | AK088795.1 | - | 2, 3 | 427 | ||
AK142391.1 | - | 2, 3 | 399 | |||
AK154895.1 | - | 2, 3 | 364 | |||
AY338464.1 | - | 2, 3 | 361 |