Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289613.1 | NP_001276542.1 | 4 | 893 | ||
NM_001289616.1 | NP_001276545.1 | 5 | 375 | |||
NM_001289620.1 | NP_001276549.1 | 5 | 375 | |||
NM_172434.3 | NP_766022.1 | 4 | 890 | |||
GenBank mRNA | AK019578.1 | - | 2 | 602 | ||
AK030259.1 | - | 3 | 201 | |||
AK044069.1 | - | 4 | 323 | |||
AK044506.1 | - | 5 | 339 | |||
AK087536.1 | - | 5 | 335 | |||
AY165052.1 | - | 4 | 608 | |||
BC031740.1 | - | 1 | 54 | |||
BC057553.1 | - | 4 | 948 | 6401356 |