Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114611.1 | NP_001108083.1 | 2 | 315 | ||
NM_001114612.1 | NP_001108084.1 | 2 | 315 | |||
NM_001114613.1 | NP_001108085.1 | 2 | 315 | |||
NM_172440.3 | NP_766028.2 | 2 | 315 | |||
GenBank mRNA | AK035159.1 | - | 2 | 281 | ||
AK043658.1 | - | 2 | 304 | |||
AK082159.1 | - | 2 | 281 | |||
AK137325.1 | - | 2 | 281 | |||
AK161955.1 | - | 2 | 288 | |||
AY542324.1 | - | 1 | 166 | |||
AY542325.1 | - | 1 | 166 | |||
AY542326.1 | - | 1 | 166 | |||
AY542327.1 | - | 1 | 166 | |||
BC137883.1 | - | 2 | 187 | 8860902 | ||
BC145007.1 | - | 2 | 187 | 9053555 | ||
BC145008.1 | - | 2 | 187 | 9053556 | ||
BC145009.1 | - | 2 | 187 | 9053557 |