Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3hav1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172467.3 | NP_766055.1 | 3 | 562 | ||
GenBank mRNA | AK045287.1 | - | 3 | 563 | ||
AK051064.1 | - | 3 | 564 | |||
AK084475.1 | - | 3 | 494 | |||
BC138180.1 | - | 3 | 654 | 8861199 | ||
BC138181.1 | - | 3 | 654 | 8861200 |