Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159527.1 | NP_001152999.1 | 2 | 265 | ||
NM_172470.3 | NP_766058.3 | 2 | 265 | |||
GenBank mRNA | AK015490.1 | - | 2 | 230 | ||
AK029896.1 | - | 2 | 220 | |||
AK031061.1 | - | 2 | 256 | |||
AK053724.1 | - | 2 | 220 | |||
AK083971.1 | - | 2 | 210 | |||
AK173151.1 | - | 2 | 207 | |||
BC056925.1 | - | 2 | 374 | 5704780 |