Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172476.4 | NP_766064.2 | 2, 3 | 332 | ||
GenBank mRNA | AK082562.1 | - | 2, 3 | 361 | ||
AK083189.1 | - | 2, 3 | 335 | |||
AY236499.1 | - | 2, 3 | 393 | |||
AY263161.1 | - | 2, 3 | 361 |