Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289699.1 | NP_001276628.1 | 2 | 757 | ||
NM_001289700.1 | NP_001276629.1 | 2 | 678 | |||
NM_172487.4 | NP_766075.2 | 2 | 731 | |||
GenBank mRNA | AK031947.1 | - | 2 | 681 | ||
AK045491.1 | - | 2 | 635 | |||
AK045678.1 | - | 2 | 719 | |||
AK136854.1 | - | 2 | 711 | |||
BC058349.1 | - | 2 | 731 | 5695444 |