Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290654.1 | NP_001277583.1 | 9 | 1927 | ||
NM_001290655.1 | NP_001277584.1 | 9 | 1951 | |||
NM_172391.4 | NP_765979.3 | 9 | 1932 | |||
GenBank mRNA | AK035941.1 | - | 9 | 1705 | ||
AK093945.1 | - | 9 | 1839 | |||
AK137116.1 | - | 9 | 1843 | |||
BC038397.1 | - | 9 | 1818 | 3676293 | ||
BC064012.1 | - | 9 | 1752 | 30135289 |