Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083331.2 | NP_001076800.1 | 17 | 2359 | ||
NM_001276406.1 | NP_001263335.1 | 17 | 2341 | |||
NM_172394.3 | NP_765982.2 | 17 | 2326 | |||
GenBank mRNA | AK002643.1 | - | 17 | 2328 | ||
AK028563.1 | - | 17 | 2349 | |||
AK050893.1 | - | 13 | 2877 | |||
AK160619.1 | - | 17 | 2327 | |||
AK166214.1 | - | 17 | 2331 | |||
BB442284.2 | - | 2 | 368 | |||
BC032929.1 | - | 17 | 2307 | 1379673 | ||
BC053390.1 | - | 2 | 2635 | |||
U01135.1 | - | 5 | 548 |