Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smim7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172396.3 | NP_765984.1 | 5 | 2046 | ||
GenBank mRNA | AF140607.1 | - | 5 | 2045 | ||
AK018610.1 | - | 5 | 2059 | |||
AK035786.1 | - | 5 | 2047 | |||
AK148562.1 | - | 5 | 2059 | |||
AK149156.1 | - | 5 | 2058 | |||
AK151539.1 | - | 5 | 2047 | |||
AK152451.1 | - | 5 | 2047 | |||
AK152748.1 | - | 5 | 2047 | |||
AK169625.1 | - | 5 | 2058 | |||
AK186598.1 | - | 1 | 159 | |||
AK207517.1 | - | 1 | 176 | |||
AK215124.1 | - | 1 | 158 | |||
AK217818.1 | - | 1 | 160 | |||
AK218215.1 | - | 1 | 159 | |||
BC055692.1 | - | 5 | 2047 | 6404961 | ||
BC060963.1 | - | 5 | 2064 |