Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172399.3 | NP_765987.2 | 4 | 4233 | ||
GenBank mRNA | AK020942.1 | - | 1 | 798 | ||
AK029210.1 | - | 4 | 4234 |