Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167767.1 | NP_001161239.1 | 6 | 1800 | ||
NM_172401.4 | NP_765989.3 | 7 | 1891 | |||
GenBank mRNA | AK004285.1 | - | 1 | 477 | ||
AK076460.1 | - | 5 | 1557 | |||
AK079099.1 | - | 7 | 1875 | |||
BC046986.1 | - | 6 | 1810 | 6476504 | ||
BC057311.1 | - | 6 | 1780 | 6811681 |