Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn2aip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172407.3 | NP_765995.1 | 3 | 3042 | ||
GenBank mRNA | AK048437.1 | - | 3 | 3014 | ||
AK078329.1 | - | 3 | 3009 | |||
AK089057.1 | - | 3 | 3011 |