Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sphkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172430.3 | NP_766018.3 | 7 | 2633 | ||
GenBank mRNA | AK034183.1 | - | 8 | 2676 | ||
AK038638.1 | - | 8 | 2678 | |||
AK122538.1 | - | 1 | 1583 | |||
BC060217.1 | - | 1 | 139 | |||
BC069832.1 | - | 7 | 2599 | 30431760 |