Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172439.3 | NP_766027.2 | 13 | 3338 | ||
GenBank mRNA | AK034272.1 | - | 13 | 3339 | ||
AK136908.1 | - | 13 | 3327 | |||
AK170114.1 | - | 13 | 3304 | |||
AK171029.1 | - | 13 | 3299 | |||
BC022915.1 | - | 13 | 2699 | |||
BC046561.1 | - | 12 | 2246 | |||
BC051452.1 | - | 13 | 3307 | |||
BY629401.1 | - | 1 | 263 |