Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035159.1 | - | 23 | 3125 | ||
BC137883.1 | - | 23 | 3031 | 8860902 | ||
BC145007.1 | - | 24 | 3025 | 9053555 | ||
BC145008.1 | - | 24 | 3103 | 9053556 | ||
BC145009.1 | - | 24 | 3124 | 9053557 |