Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039388.2 | NP_001034477.1 | 14 | 2418 | ||
NM_172445.3 | NP_766033.1 | 14 | 2339 | |||
GenBank mRNA | AK032582.1 | - | 14 | 2301 | ||
AK036203.1 | - | 14 | 2335 | |||
AK129254.1 | - | 14 | 2295 | |||
AK146209.1 | - | 14 | 2314 | |||
BC046236.1 | - | 13 | 2166 | 6817089 | ||
BC046562.1 | - | 3 | 899 |