Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161767.1 | NP_001155239.1 | 3 | 798 | ||
NM_001161768.1 | NP_001155240.1 | 3 | 795 | |||
NM_172451.3 | NP_766039.2 | 3 | 794 | |||
GenBank mRNA | AJ133523.1 | - | 1 | 407 | ||
AK028506.1 | - | 3 | 794 | |||
AK049222.1 | - | 3 | 797 | |||
AK155008.1 | - | 3 | 801 | |||
AK159721.1 | - | 3 | 776 | |||
BC119324.1 | - | 1 | 447 | 8734073 | ||
BC119326.1 | - | 1 | 447 | 8734075 |