Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105196.1 | NP_001098666.1 | 9 | 2897 | ||
NM_001105197.1 | NP_001098667.1 | 10 | 2688 | |||
NM_001271489.1 | NP_001258418.1 | 10 | 2983 | |||
NM_001271490.1 | NP_001258419.1 | 10 | 3155 | |||
NM_001271491.1 | NP_001258420.1 | 9 | 2617 | |||
NM_172472.3 | NP_766060.2 | 10 | 3002 | |||
GenBank mRNA | AK089762.1 | - | 8 | 2659 | ||
AK139756.1 | - | 9 | 2894 | |||
AK144657.1 | - | 10 | 2967 | |||
AK144751.1 | - | 10 | 2974 | |||
AK145029.1 | - | 10 | 3124 | |||
AK145832.1 | - | 10 | 2968 | |||
AK147405.1 | - | 7 | 6061 | |||
AK150372.1 | - | 10 | 2754 | |||
AK151447.1 | - | 10 | 2673 | |||
AK152221.1 | - | 10 | 2673 | |||
AK153051.1 | - | 10 | 2754 | |||
AK154690.1 | - | 8 | 4456 | |||
AK155883.1 | - | 9 | 2619 | |||
AK159162.1 | - | 10 | 2882 | |||
BC056358.1 | - | 10 | 2990 | 6813486 | ||
BC063047.1 | - | 10 | 2990 | 6813486 | ||
S76673.1 | - | 8 | 2340 |