Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172477.4 | NP_766065.1 | 19 | 3854 | ||
GenBank mRNA | AK048041.1 | - | 19 | 3839 | ||
AK081176.1 | - | 19 | 3854 | |||
AK135781.1 | - | 17 | 2399 | |||
AK160784.1 | - | 19 | 3616 | |||
AK165202.1 | - | 19 | 3709 | |||
BC056367.1 | - | 19 | 3724 | |||
BC057310.1 | - | 19 | 3724 |