Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nckap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081756.1 | NP_001075225.1 | 17 | 5979 | ||
NM_172484.3 | NP_766072.2 | 15 | 5840 | |||
NM_176957.3 | NP_795931.2 | 13 | 1732 | |||
GenBank mRNA | AK053243.1 | - | 11 | 1415 | ||
AY957601.1 | - | 12 | 5207 | |||
AY957602.1 | - | 13 | 5399 | |||
BC066095.1 | - | 15 | 5840 | 6856184 | ||
BC079551.1 | - | 15 | 5840 | 6856184 |