Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145001.2 | NP_001138473.1 | 11 | 2220 | ||
NM_001166167.1 | NP_001159639.1 | 10 | 2002 | |||
NM_001166168.1 | NP_001159640.1 | 10 | 1997 | |||
NM_001166169.1 | NP_001159641.1 | 10 | 2006 | |||
NM_001166170.1 | NP_001159642.1 | 10 | 2004 | |||
NM_001166171.1 | NP_001159643.1 | 11 | 2169 | |||
NM_014397.5 | NP_055212.2 | 10 | 2055 | |||
XM_005251664.1 | XP_005251721.1 | 10 | 2061 | |||
XM_006716936.2 | XP_006716999.1 | 10 | 2066 | |||
XM_017014217.1 | XP_016869706.1 | 11 | 2229 | |||
GenBank mRNA | AJ420460.1 | - | 1 | 260 | ||
BC000101.2 | - | 10 | 2001 | 3507262 | ||
BC012761.2 | - | 10 | 2002 | 3640103 | ||
BP219389.1 | - | 1 | 564 |