Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172514.3 | NP_766102.1 | 4 | 599 | ||
GenBank mRNA | AK088711.1 | - | 4 | 600 | ||
AK170268.1 | - | 4 | 596 | |||
BC117823.1 | - | 4 | 532 | 40092863 | ||
BC117824.1 | - | 4 | 532 | 40092864 |