Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnksr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172546.2 | NP_766134.1 | 2 | 754 | ||
GenBank mRNA | AK033015.1 | - | 2 | 755 | ||
AK149951.1 | - | 1 | 53 | |||
AY151137.1 | - | 2 | 716 | |||
BC024086.1 | - | 1 | 102 |