Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166669.1 | NP_001160141.1 | 2 | 370 | ||
NM_001166670.1 | NP_001160142.1 | 2 | 370 | |||
NM_001166671.1 | NP_001160143.1 | 2 | 370 | |||
NM_172558.3 | NP_766146.2 | 2 | 370 | |||
GenBank mRNA | AK049216.1 | - | 2 | 305 | ||
AK131137.1 | - | 2 | 346 | |||
AK143536.1 | - | 2 | 319 | |||
BC137802.1 | - | 2 | 313 | 8860821 | ||
BC144921.1 | - | 2 | 310 | |||
BC144922.1 | - | 2 | 313 | 9053470 | ||
BY742908.1 | - | 2 | 371 |